6-(Ethylsulphonyl)-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Ethylsulphonyl)-4H-benzo[b][1,4]oxazin-3-one, 3,4-Dihydro-3-oxo-2H-1,4-benzoxazin-6-yl ethyl sulphone

OR9456-100MG
APOSOR9456-100MGEA 29 EUR
APOSOR9456-100MG
6-(Ethylsulphonyl)-2H-1,4-benzoxazin-3(4H)-one
6-(Ethylsulphonyl)-2H-1,4-benzoxazin-3(4H)-one
Formel: C₁₀H₁₁NO₄S
Molecular Weight: 241,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00798368
CAS-Nummer: 874774-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR