6-Chloro-N-isopropylpyridazin-3-amine 95%

Lieferant: Apollo Scientific

OR945691-1G
APOSOR945691-1GEA 92.8 EUR
APOSOR945691-1G
6-Chloro-N-isopropylpyridazin-3-amine 95%
6-Chloro-N-isopropylpyridazin-3-amine
Formel: C₇H₁₀ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11125896
CAS-Nummer: 1007-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR