N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine 95%
Lieferant: Apollo Scientific
OR945862-1G
APOSOR945862-1GEA
163
EUR
APOSOR945862-1G
N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine 95%
N,N-Dimethyl-1H-pyrazolo[3,4-b]pyridin-6-amine
Formel:
C₈H₁₀N₄ Molecular Weight: 162.192 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13178167 CAS-Nummer: 63725-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...