5-Methyl-2-thenoesäure

Lieferant: Apollo Scientific

Synonyme: 5-Methylthiophene-2-carboxylic acid

OR9459-100G OR9459-25G OR9459-5G
APOSOR9459-100GEA 110 EUR
APOSOR9459-100G APOSOR9459-25G APOSOR9459-5G
5-Methyl-2-thenoesäure
5-Methyl-2-thenoesäure
Formel: C₆H₆O₂S
Schmelzpunkt: 135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005439
CAS-Nummer: 1918-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR