5-Methyl-2-thenoesäure
Lieferant: Apollo Scientific
Synonyme:
5-Methylthiophene-2-carboxylic acid
OR9459-100G
OR9459-25G
OR9459-5G
APOSOR9459-100GEA
110
EUR
APOSOR9459-100G
APOSOR9459-25G
APOSOR9459-5G
5-Methyl-2-thenoesäure
5-Methyl-2-thenoesäure
Formel:
C₆H₆O₂S Schmelzpunkt: 135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005439 CAS-Nummer: 1918-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...