1,2-Cyclopentane diformic anhydride 98%
Lieferant: Apollo Scientific
Synonyme:
Tetrahydro-1H-cyclopenta[c]furan-1,3(3ah)-dione
OR945919-1G
OR945919-5G
APOSOR945919-1GEA
65.4
EUR
APOSOR945919-1G
APOSOR945919-5G
1,2-Cyclopentane diformic anhydride 98%
1,2-Cyclopentane diformic anhydride
Formel:
C₇H₈O₃ Molecular Weight: 140.138 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
5763-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...