5-Methyl-D-tryptophan 97%

Lieferant: Apollo Scientific

OR946064-1G OR946064-250MG OR946064-500MG
APOSOR946064-1GEA 870 EUR
APOSOR946064-1G APOSOR946064-250MG APOSOR946064-500MG
5-Methyl-D-tryptophan 97%
5-Methyl-D-tryptophan
Formel: C₁₂H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09841957
CAS-Nummer: 99295-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR