5-Methyl-D-tryptophan 97%
Lieferant: Apollo Scientific
OR946064-1G
OR946064-250MG
OR946064-500MG
APOSOR946064-1GEA
870
EUR
APOSOR946064-1G
APOSOR946064-250MG
APOSOR946064-500MG
5-Methyl-D-tryptophan 97%
5-Methyl-D-tryptophan
Formel:
C₁₂H₁₄N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09841957 CAS-Nummer: 99295-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...