1,7-Diazaspiro[3.5]nonan-2-one
Lieferant: Apollo Scientific
OR946098-1G
APOSOR946098-1GEA
427
EUR
APOSOR946098-1G
1,7-Diazaspiro[3.5]nonan-2-one
1,7-Diazaspiro[3.5]nonan-2-one
Formel:
C₇H₁₂N₂O Molecular Weight: 140,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14705692 CAS-Nummer: 1235440-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...