3-[(4-Oxo-3,4-dihydro-1-phthalazinyl)methyl]benzoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-((4-Oxo-3,4-dihydrophthalazin-1-yl)methyl)benzoic acid

OR946199-250MG
APOSOR946199-250MGEA 157 EUR
APOSOR946199-250MG
3-[(4-Oxo-3,4-dihydro-1-phthalazinyl)methyl]benzoic acid 95%
3-[(4-Oxo-3,4-dihydro-1-phthalazinyl)methyl]benzoic acid
Formel: C₁₆H₁₂N₂O₃
Molecular Weight: 280,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20483902
CAS-Nummer: 420846-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR