4-Isopropyl-5-pyrimidinboronsäure 96%

Lieferant: Apollo Scientific
Achtung

OR9462-25MG
APOSOR9462-25MGEA 31.9 EUR
APOSOR9462-25MG
4-Isopropyl-5-pyrimidinboronsäure 96%
4-Isopropyl-5-pyrimidinboronsäure
Formel: C₇H₁₁BN₂O₂
Molecular Weight: 165.99 g/mol
Schmelzpunkt: 104…109 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235084
CAS-Nummer: 913835-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR