(S)-2-ethylpyrrolidine-2-carboxylic acid hydrochloride ≥98%

Lieferant: Apollo Scientific

OR946257-100MG
APOSOR946257-100MGEA 323 EUR
APOSOR946257-100MG
(S)-2-ethylpyrrolidine-2-carboxylic acid hydrochloride ≥98%
(S)-2-ethylpyrrolidine-2-carboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20487711
CAS-Nummer: 1431699-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR