2-Chlor-5-ethoxyphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chloro-5-ethoxyphenyl)boronic acid

OR9463-1G
APOSOR9463-1GEA 52.2 EUR
APOSOR9463-1G
2-Chlor-5-ethoxyphenylboronsäure 97%
2-Chlor-5-ethoxyphenylboronsäure
Formel: C₈H₁₀BClO₃
Molecular Weight: 200,427 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235081
CAS-Nummer: 913835-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR