5-(1-Hydroxyethyl)-2-methylpyridine 95%

Lieferant: Apollo Scientific

OR946325-1G
APOSOR946325-1GEA 126 EUR
APOSOR946325-1G
5-(1-Hydroxyethyl)-2-methylpyridine 95%
5-(1-Hydroxyethyl)-2-methylpyridine
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06637422
CAS-Nummer: 100189-16-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR