4-(Chloromethyl)-N,N-dimethylbenzamide 95%

Lieferant: Apollo Scientific

OR946362-1G
APOSOR946362-1GEA 806 EUR
APOSOR946362-1G
4-(Chloromethyl)-N,N-dimethylbenzamide 95%
4-(Chloromethyl)-N,N-dimethylbenzamide
Formel: C₁₀H₁₂ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11204892
CAS-Nummer: 121083-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR