3-Iodo-N,N-dimethylbenzamide 98%

Lieferant: Apollo Scientific
Achtung

OR946415-1G
APOSOR946415-1GEA 58 EUR
APOSOR946415-1G
3-Iodo-N,N-dimethylbenzamide 98%
3-Iodo-N,N-dimethylbenzamide
Formel: C₉H₁₀INO
Lagertemperatur: Raumtemperatur
MDL: MFCD02859819
CAS-Nummer: 91506-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR