2-(4-Bromophenoxy)benzonitrile 98%

Lieferant: Apollo Scientific
Achtung

OR946471-1G
APOSOR946471-1GEA 400 EUR
APOSOR946471-1G
2-(4-Bromophenoxy)benzonitrile 98%
2-(4-Bromophenoxy)benzonitrile
Formel: C₁₃H₈BrNO
Molecular Weight: 274,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10564769
CAS-Nummer: 330793-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR