5-(2-Chloroethyl)-2,3-dihydrobenzofuran 95%

Lieferant: Apollo Scientific

OR946504-1G
APOSOR946504-1GEA 348 EUR
APOSOR946504-1G
5-(2-Chloroethyl)-2,3-dihydrobenzofuran 95%
5-(2-Chloroethyl)-2,3-dihydrobenzofuran
Formel: C₁₀H₁₁ClO
Molecular Weight: 182,647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11046521
CAS-Nummer: 943034-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR