3-(2-Methylphenoxy)azetidine 95%

Lieferant: Apollo Scientific

Synonyme: 3-(2-Methylphenoxy)azetidine hydrochloride

OR946592-1G
APOSOR946592-1GEA 464 EUR
APOSOR946592-1G
3-(2-Methylphenoxy)azetidine 95%
3-(2-Methylphenoxy)azetidine
Formel: C₁₀H₁₃NO
Molecular Weight: 163,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09861857
CAS-Nummer: 954222-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR