3-(3-Methylphenoxy)azetidine 95%

Lieferant: Apollo Scientific

OR946593-1G
APOSOR946593-1GEA 464 EUR
APOSOR946593-1G
3-(3-Methylphenoxy)azetidine 95%
3-(3-Methylphenoxy)azetidine
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09861892
CAS-Nummer: 949100-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR