Ethyl 1-methyl-4-oxopiperidine-3-carboxylate 95%

Lieferant: Apollo Scientific

OR946681-1G
APOSOR946681-1GEA 609 EUR
APOSOR946681-1G
Ethyl 1-methyl-4-oxopiperidine-3-carboxylate 95%
Ethyl 1-methyl-4-oxopiperidine-3-carboxylate
Formel: C₉H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00627739
CAS-Nummer: 25012-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR