2-Bromo-6-methylisonicotinic acid ≥98%

Lieferant: Apollo Scientific

OR946687-250MG
APOSOR946687-250MGEA 97.3 EUR
APOSOR946687-250MG
2-Bromo-6-methylisonicotinic acid ≥98%
2-Bromo-6-methylisonicotinic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,032 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13188656
CAS-Nummer: 25462-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR