Octa-3,5-diyne-1,8-diol 95%

Lieferant: Apollo Scientific

OR946829-1G
APOSOR946829-1GEA 647 EUR
APOSOR946829-1G
Octa-3,5-diyne-1,8-diol 95%
Octa-3,5-diyne-1,8-diol
Formel: C₈H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00190210
CAS-Nummer: 15808-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR