2-(N-Benzyl-N-methylamino)acetonitrile 95%
Lieferant: Apollo Scientific
OR946881-1G
APOSOR946881-1GEA
145
EUR
APOSOR946881-1G
2-(N-Benzyl-N-methylamino)acetonitrile 95%
2-(N-Benzyl-N-methylamino)acetonitrile
Formel:
C₁₀H₁₂N₂ Molecular Weight: 160,216 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09906884 CAS-Nummer: 14321-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...