tert-Butyl diprop-2-ynylcarbamate 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl di(prop-2-yn-1-yl)carbamate

OR946951-1G
APOSOR946951-1GEA 69.6 EUR
APOSOR946951-1G
tert-Butyl diprop-2-ynylcarbamate 95%
tert-Butyl diprop-2-ynylcarbamate
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16621687
CAS-Nummer: 262418-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR