tert-Butyl-4-methoxycarbanilate 95%

Lieferant: Apollo Scientific

OR946958-5G
APOSOR946958-5GEA 21.9 EUR
APOSOR946958-5G
tert-Butyl-4-methoxycarbanilate 95%
tert-Butyl-4-methoxycarbanilate
Formel: C₁₂H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00816766
CAS-Nummer: 18437-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR