2,2-Dimethyl-3-oxobutanoic acid 95%

Lieferant: Apollo Scientific

OR946995-250MG
APOSOR946995-250MGEA 522 EUR
APOSOR946995-250MG
2,2-Dimethyl-3-oxobutanoic acid 95%
2,2-Dimethyl-3-oxobutanoic acid
Formel: C₆H₁₀O₃
Molecular Weight: 130,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16620293
CAS-Nummer: 98485-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR