2-Methyl-N-((oxiran-2-yl)methyl)propan-2-amine 95%
Lieferant: Apollo Scientific
OR947056-1G
APOSOR947056-1GEA
85.7
EUR
APOSOR947056-1G
2-Methyl-N-((oxiran-2-yl)methyl)propan-2-amine 95%
2-Methyl-N-((oxiran-2-yl)methyl)propan-2-amine
Formel:
C₇H₁₅NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD16620337 CAS-Nummer: 13080-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...