Methyl 2-oxo-2-(1H-pyrrol-2-yl)acetate 95%

Lieferant: Apollo Scientific

OR947057-1G
APOSOR947057-1GEA 308 EUR
APOSOR947057-1G
Methyl 2-oxo-2-(1H-pyrrol-2-yl)acetate 95%
Methyl 2-oxo-2-(1H-pyrrol-2-yl)acetate
Formel: C₇H₇NO₃
Molecular Weight: 153,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16620338
CAS-Nummer: 54224-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR