3-Oxo-3-(3-tolyl)propionic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-oxo-3-(m-tolyl)propanoate

OR947058-1G
APOSOR947058-1GEA 108 EUR
APOSOR947058-1G
3-Oxo-3-(3-tolyl)propionic acid methyl ester 95%
3-Oxo-3-(3-tolyl)propionic acid methyl ester
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086702
CAS-Nummer: 200404-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR