3-Oxo-3-(2-tolyl)propionic acid methyl ester 95%

Lieferant: Apollo Scientific

OR947059-1G
APOSOR947059-1GEA 400 EUR
APOSOR947059-1G
3-Oxo-3-(2-tolyl)propionic acid methyl ester 95%
3-Oxo-3-(2-tolyl)propionic acid methyl ester
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424779
CAS-Nummer: 147501-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR