Methyl-3-(4-methylphenyl)-3-oxopropanoate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-oxo-3-(p-tolyl)propanoate

OR947060-1G
APOSOR947060-1GEA 154 EUR
APOSOR947060-1G
Methyl-3-(4-methylphenyl)-3-oxopropanoate 95%
Methyl-3-(4-methylphenyl)-3-oxopropanoate
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424778
CAS-Nummer: 22027-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR