4-(3-Hydroxypropylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: (4-((3-Hydroxypropyl)carbamoyl)phenyl)boronic acid

OR9471-1G
APOSOR9471-1GEA 142 EUR
APOSOR9471-1G
4-(3-Hydroxypropylcarbamoyl)phenylboronsäure 97%
4-(3-Hydroxypropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₄
Molecular Weight: 223,0335 g/mol
Schmelzpunkt: 224…226 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235076
CAS-Nummer: 913835-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR