N1,N2-Diethylbenzene-1,2-diamine 95%
Lieferant: Apollo Scientific
OR947185-1G
APOSOR947185-1GEA
197
EUR
APOSOR947185-1G
N1,N2-Diethylbenzene-1,2-diamine 95%
N1,N2-Diethylbenzene-1,2-diamine
Formel:
C₁₀H₁₆N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14582999 CAS-Nummer: 24340-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...