N1,N3-Diethylbenzene-1,3-diamine 95%

Lieferant: Apollo Scientific

OR947186-1G
APOSOR947186-1GEA 168 EUR
APOSOR947186-1G
N1,N3-Diethylbenzene-1,3-diamine 95%
N1,N3-Diethylbenzene-1,3-diamine
Formel: C₁₀H₁₆N₂
Molecular Weight: 164,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583000
CAS-Nummer: 5857-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR