(E)-Methyl 2-cyano-3-ethoxyacrylate 95%

Lieferant: Apollo Scientific

OR947242-1G OR947242-25G OR947242-5G
APOSOR947242-1GEA 125 EUR
APOSOR947242-1G APOSOR947242-25G APOSOR947242-5G
(E)-Methyl 2-cyano-3-ethoxyacrylate 95%
(E)-Methyl 2-cyano-3-ethoxyacrylate
Formel: C₇H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00087470
CAS-Nummer: 71363-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR