2-(2-Nitrophenyl)-1,3-dioxolane 95%

Lieferant: Apollo Scientific

OR947260-1G
APOSOR947260-1GEA 223 EUR
APOSOR947260-1G
2-(2-Nitrophenyl)-1,3-dioxolane 95%
2-(2-Nitrophenyl)-1,3-dioxolane
Formel: C₉H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06209042
CAS-Nummer: 48140-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR