N,N'-(1,3-Phenylene)diacetamide 95%

Lieferant: Apollo Scientific

OR947264-1G
APOSOR947264-1GEA 294 EUR
APOSOR947264-1G
N,N'-(1,3-Phenylene)diacetamide 95%
N,N'-(1,3-Phenylene)diacetamide
Formel: C₁₀H₁₂N₂O₂
Molecular Weight: 192.214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035792
CAS-Nummer: 10268-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR