3-(2,2,2-Trichloroacetamido)benzeneboronic acid 97%
Lieferant: Apollo Scientific
OR9473-1G
APOSOR9473-1GEA
176
EUR
APOSOR9473-1G
3-(2,2,2-Trichloroacetamido)benzeneboronic acid 97%
3-(2,2,2-Trichloroacetamido)benzeneboronic acid
Formel:
C₈H₇BCl₃NO₃ Molecular Weight: 282.316 g/mol Schmelzpunkt: 220…222 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235073 CAS-Nummer: 276669-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...