2-(2-Cyano-4-methoxyphenyl)acetic acid 95%
Lieferant: Apollo Scientific
OR947312-1G
OR947312-250MG
APOSOR947312-1GEA
446
EUR
APOSOR947312-1G
APOSOR947312-250MG
2-(2-Cyano-4-methoxyphenyl)acetic acid 95%
2-(2-Cyano-4-methoxyphenyl)acetic acid
Formel:
C₁₀H₉NO₃ Molecular Weight: 191.183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18394271 CAS-Nummer: 52786-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...