2-Chloro-4-cyanobenzyl alcohol 95%

Lieferant: Apollo Scientific

OR947344-1G
APOSOR947344-1GEA 560 EUR
APOSOR947344-1G
2-Chloro-4-cyanobenzyl alcohol 95%
2-Chloro-4-cyanobenzyl alcohol
Formel: C₈H₆ClNO
Molecular Weight: 167.592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18392443
CAS-Nummer: 1261618-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR