2,4-Dicyananilin 95%

Lieferant: Apollo Scientific

Synonyme: 4-Aminoisophthalonitrile

OR947348-1G
APOSOR947348-1GEA 62.5 EUR
APOSOR947348-1G
2,4-Dicyananilin 95%
2,4-Dicyananilin
Formel: C₈H₅N₃
Molecular Weight: 143.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270798
CAS-Nummer: 19619-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR