2-(2-Methoxy-3-methylphenyl)acetic acid 95%

Lieferant: Apollo Scientific

OR947378-1G
APOSOR947378-1GEA 400 EUR
APOSOR947378-1G
2-(2-Methoxy-3-methylphenyl)acetic acid 95%
2-(2-Methoxy-3-methylphenyl)acetic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398339
CAS-Nummer: 854812-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR