3H-Imidazo[4,5-c]pyridin-2-ol 95%

Lieferant: Apollo Scientific

OR947396-5G
APOSOR947396-5GEA 154 EUR
APOSOR947396-5G
3H-Imidazo[4,5-c]pyridin-2-ol 95%
3H-Imidazo[4,5-c]pyridin-2-ol
Formel: C₆H₅N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18872694
CAS-Nummer: 7397-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR