Methyl-2-methoxyisonicotinat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-methoxy-4-pyridinecarboxylate, Methyl 2-methoxyisonicotinate

OR9474-250MG
APOSOR9474-250MGEA 21.9 EUR
APOSOR9474-250MG
Methyl-2-methoxyisonicotinat 97%
Methyl-2-methoxyisonicotinat
Formel: C₈H₉NO₃
Molecular Weight: 167,1620 g/mol
Schmelzpunkt: 33…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08436071
CAS-Nummer: 26156-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR