6-Nitro-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR9475-25G
APOSOR9475-25GEA
24.8
EUR
APOSOR9475-25G
6-Nitro-1H-indazol
6-Nitro-1H-indazole
Formel:
C₇H₅N₃O₂ Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005695 CAS-Nummer: 7597-18-4 EINECS: 231-500-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...