2,6-Diazaspiro[3.4]octan-7-one 95%

Lieferant: Apollo Scientific
Achtung

OR947562-250MG
APOSOR947562-250MGEA 720 EUR
APOSOR947562-250MG
2,6-Diazaspiro[3.4]octan-7-one 95%
2,6-Diazaspiro[3.4]octan-7-one
Formel: C₆H₁₀N₂O
Molecular Weight: 126,156 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21364482
CAS-Nummer: 1211515-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR