N-Formyl-L-phenylalanine 95%
Lieferant: Apollo Scientific
Synonyme:
For-Phe-OH, (S)-2-Formamido-3-phenylpropanoic acid
OR947569-1G
APOSOR947569-1GEA
43.5
EUR
APOSOR947569-1G
N-Formyl-L-phenylalanine 95%
N-Formyl-L-phenylalanine
Formel:
C₁₀H₁₁NO₃ Molecular Weight: 193.20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021030 CAS-Nummer: 13200-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...