2-(Methoxymethyl)oxiran 90%

Lieferant: Apollo Scientific

Synonyme: 1,2-Epoxy-3-methoxypropane

OR947696-25G OR947696-5G OR947696-1G
APOSOR947696-25GEA 0 EUR
APOSOR947696-25G APOSOR947696-5G APOSOR947696-1G
2-(Methoxymethyl)oxiran 90%
Glycidyl methyl ether
Formel: C₄H₈O₂
Flammpunkt: 25 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005140
CAS-Nummer: 930-37-0
UN: 1992
ADR: 3+6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR