2-(Methoxymethyl)oxiran 90%
Lieferant: Apollo Scientific
Synonyme:
1,2-Epoxy-3-methoxypropane
OR947696-25G
OR947696-5G
OR947696-1G
APOSOR947696-25GEA
0
EUR
APOSOR947696-25G
APOSOR947696-5G
APOSOR947696-1G
2-(Methoxymethyl)oxiran 90%
Glycidyl methyl ether
Formel:
C₄H₈O₂ Flammpunkt: 25 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005140 CAS-Nummer: 930-37-0 UN: 1992 ADR: 3+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...